Product Image
Invitrogen™

IgM Mouse ELISA Standard

This product is a component of a respective eBioscience Kit. Please refer to the relevant manual for each product onRead more
Have Questions?
Catalog NumberQuantity
39-50470-654 pack
Catalog number 39-50470-65
Price (INR)
12,470.00
Online offer
Ends: 31-Mar-2026
20,783.00
Save 8,313.00 (40%)
Each
Quantity:
4 pack
Price (INR)
12,470.00
Online offer
Ends: 31-Mar-2026
20,783.00
Save 8,313.00 (40%)
Each
This product is a component of a respective eBioscience Kit. Please refer to the relevant manual for each product on our website.

Immunoglobulin M is the third most common serum Ig and can exist as both a pentamer and as a monomer (e.g. found on B lymphocytes as B - cell receptors).

The large pentameric structure allows for building of bridges between encountered epitopes on molecules that are too distant as to be connected by smaller IgG antibodies.

IgM is built as the first antibody during an immune response and is responsible for agglutination and cytolytic reactions since its pentameric structure gives it 10 free antigen binding sites in theory and it posseses a high avidity. Due to conformational constraints among the 10 Fab portions, IgM only has a valence of 5. Additionally, IgM is not as versatile as IgG, but it is of vital importance in complement activation and agglutination.

IgM is predominantly found in the lymph fluid and the blood and is a very effective neutralizing agent in the early stages of disease. Elevated levels can be a sign of recent infection or exposure to antigen.

Reported Application
ELISA, ELISA Recombinant Standard

For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Equipment)Microplate Reader
FormatKit
Former IdentifierseBioscience
Immunoassay Kit FormatELISA Standard
Quantity4 pack
Target Kit NamedIgM
Target Species NamedMouse
Target Species PrimaryMouse
Target Species ValidatedMouse
FormLiquid
RecombinantRecombinant
Unit SizeEach