N,N-Diisopropylethylamine trihydrofluoride, 90%
N,N-Diisopropylethylamine trihydrofluoride, 90%
N,N-Diisopropylethylamine trihydrofluoride, 90%
Acros Organics

N,N-Diisopropylethylamine trihydrofluoride, 90%

CAS: 131600-43-6 | C8H22F3N | 189.27 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Packaging:
Plastic bottle
Catalog number 390710050
also known as 39071
HSN 29211990
Price (INR)
36,055.00
Each
Quantity:
5 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
36,055.00
Each
Chemical Identifiers
CAS131600-43-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to yellow-brown
Appearance (Form)Liquid
Titration with NaOH18 to 22 % (available HF)
Infrared spectrumConforms
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only