Xylenes, extra pure, mixture of isomers
Xylenes, extra pure, mixture of isomers
Xylenes, extra pure, mixture of isomers
Xylenes, extra pure, mixture of isomers
Acros Organics

Xylenes, extra pure, mixture of isomers

CAS: 1330-20-7 | C8H10
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
2.5 L
Packaging:
Glass Bottle
Catalog number 390790010
also known as 39079
HSN 29024400
Price (INR)
5,326.00
Each
Add to cart
Quantity:
1 L
Packaging:
Glass Bottle
Request bulk or custom format
Price (INR)
5,326.00
Each
Add to cart
Chemical Identifiers
CAS1330-20-7
IUPAC Name1,3-xylene
Molecular FormulaC8H10
InChI KeyIVSZLXZYQVIEFR-UHFFFAOYSA-N
SMILESCC1=CC(C)=CC=C1
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=98.0 % (mixed isomers)
Substance darkened by H2SO4Passes test
Appearance (Color)Clear colorless
Appearance (Form)Liquid
Water=<0.05 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only