4,4'-Biphenyldicarboxaldehyde, 97%
4,4'-Biphenyldicarboxaldehyde, 97%
4,4'-Biphenyldicarboxaldehyde, 97%
Acros Organics

4,4'-Biphenyldicarboxaldehyde, 97%

CAS: 66-98-8 | C14H10O2 | 210.23 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 391130010
also known as 39113
HSN 29122990
Price (INR)
9,279.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,279.00
Each
Chemical Identifiers
CAS66-98-8
IUPAC Name[1,1'-biphenyl]-4,4'-dicarbaldehyde
Molecular FormulaC14H10O2
InChI KeyFEHLIYXNTWAEBQ-UHFFFAOYSA-N
SMILESO=CC1=CC=C(C=C1)C1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Melting point144°C to 147°C
Appearance (Form)Crystalline powder
Appearance (Color)Yellow
Infrared spectrumConforms
GC>=96.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only