1,2-Bis(2-iodoethoxy)ethane, 97%, stabilized
1,2-Bis(2-iodoethoxy)ethane, 97%, stabilized
1,2-Bis(2-iodoethoxy)ethane, 97%, stabilized
Acros Organics

1,2-Bis(2-iodoethoxy)ethane, 97%, stabilized

CAS: 36839-55-1 | C6H12I2O2 | 369.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
25 g
Packaging:
Glass bottle
Catalog number 391690050
also known as 39169
HSN 29091900
Price (INR)
7,078.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
7,078.00
Each
Chemical Identifiers
CAS36839-55-1
IUPAC Name1-iodo-2-[2-(2-iodoethoxy)ethoxy]ethane
Molecular FormulaC6H12I2O2
InChI KeyBCAGFJXMCZSAHD-UHFFFAOYSA-N
SMILESICCOCCOCCI
View more
SpecificationsSpecification SheetSpecification Sheet
GC>=96.0 %
Appearance (Form)Liquid
Infrared spectrumConforms
Appearance (Color)Clear yellow to orange-red
Stabilizercopper
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only