Phenyl propargyl ether, 97+%
Phenyl propargyl ether, 97+%
Phenyl propargyl ether, 97+%
Acros Organics

Phenyl propargyl ether, 97+%

CAS: 13610-02-1 | C9H8O | 132.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 mL
5 mL
Packaging:
Glass bottle
Catalog number 392730250
also known as 39273
HSN 29093090
Price (INR)
37,117.00
Each
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
37,117.00
Each
Chemical Identifiers
CAS13610-02-1
SpecificationsSpecification SheetSpecification Sheet
Refractive index1.5320 to 1.5350 (20°C, 589 nm)
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=97.0 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only