Diphosphoryl chloride, 90+%
Diphosphoryl chloride, 90+%
Diphosphoryl chloride, 90+%
Diphosphoryl chloride, 90+%
Acros Organics

Diphosphoryl chloride, 90+%

CAS: 13498-14-1 | Cl4O3P2 | 251.76 g/mol
Have Questions?
Catalog NumberQuantityPackaging
393620250
also known as 39362
25 mLGlass bottle
Catalog number 393620250
also known as 39362
HSN 29319090
Price (INR)
29,282.00
Each
Add to cart
Quantity:
25 mL
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
29,282.00
Each
Add to cart
Chemical Identifiers
CAS13498-14-1
IUPAC Name[(dichlorophosphoryl)oxy]phosphonoyl dichloride
Molecular FormulaCl4O3P2
InChI KeyCNTIXUGILVWVHR-UHFFFAOYSA-N
SMILESClP(Cl)(=O)OP(Cl)(Cl)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Appearance (Color)Clear colorless to yellow to light orange
Titration with NaOH>=90.0 %
Appearance (Form)Liquid
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only