Methylthymol blue, tetrasodium salt, ACS reagent, indicator grade
Methylthymol blue, tetrasodium salt, ACS reagent, indicator grade
Methylthymol blue, tetrasodium salt, ACS reagent, indicator grade
Acros Organics

Methylthymol blue, tetrasodium salt, ACS reagent, indicator grade

CAS: 1945-77-3 | C37H44N2Na4O13S | 848.779 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Packaging:
Glass bottle
Catalog number 394210010
HSN 32041911
Price (INR)
5,147.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
5,147.00
Each
Chemical Identifiers
CAS1945-77-3
IUPAC Nametetrasodium 2-[({3-[3-(3-{[bis(carboxylatomethyl)amino]methyl}-4-hydroxy-2-methyl-5-(propan-2-yl)phenyl)-1,1-dioxo-3H-2,1λ⁶-benzoxathiol-3-yl]-6-hydroxy-2-methyl-5-(propan-2-yl)phenyl}methyl)(carboxylatomethyl)amino]acetate
Molecular FormulaC37H40N2Na4O13S
InChI KeyDIZZDZCUMBBRSG-UHFFFAOYSA-J
SMILES[Na+].[Na+].[Na+].[Na+].CC(C)C1=CC(=C(C)C(CN(CC([O-])=O)CC([O-])=O)=C1O)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(C(C)C)=C(O)C(CN(CC([O-])=O)CC([O-])=O)=C1C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Dark green to brown to black
Appearance (Form)Powder
Infrared spectrumConforms
Clarity of solutionPasses test
Suitability(as metal indicator) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only