N-BOC-3-pyrrolidinone, 97%
N-BOC-3-pyrrolidinone, 97%
N-BOC-3-pyrrolidinone, 97%
Acros Organics

N-BOC-3-pyrrolidinone, 97%

CAS: 101385-93-7 | C9H15NO3 | 185.22 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
10 g
Packaging:
Glass bottle
Catalog number 397050010
also known as 39705
HSN 29222990
Price (INR)
9,398.00
Each
Quantity:
1 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,398.00
Each
Chemical Identifiers
CAS101385-93-7
IUPAC Nametert-butyl 3-oxopyrrolidine-1-carboxylate
Molecular FormulaC9H15NO3
InChI KeyJSOMVCDXPUXKIC-UHFFFAOYSA-N
SMILESCC(C)(C)OC(=O)N1CCC(=O)C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light brown
Appearance (Form)Low melting solid
Infrared spectrumConforms
GC>=97.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only