3-(Bromomethyl)pyridine hydrobromide, 97%
3-(Bromomethyl)pyridine hydrobromide, 97%
3-(Bromomethyl)pyridine hydrobromide, 97%
Acros Organics

3-(Bromomethyl)pyridine hydrobromide, 97%

CAS: 4916-55-6 | C6H6BrN·HBr | 252.93 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
1 g
5 g
Catalog number 397140010
also known as 39714
HSN 29333919
Price (INR)
10,009.00
Each
Quantity:
1 g
Request bulk or custom format
Price (INR)
10,009.00
Each
Chemical Identifiers
CAS4916-55-6
IUPAC Namehydrogen 3-(bromomethyl)pyridine bromide
Molecular FormulaC6H7Br2N
InChI KeyFNHPUOJKUXFUKN-UHFFFAOYSA-N
SMILES[H+].[Br-].BrCC1=CC=CN=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder or crystals and/or chunks
Infrared spectrumConforms
Melting point148°C to 154°C
Titration with HClO4>=96.0 %
Appearance (Color)White to yellow to brown
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only