Cyclopentyl methyl ether, 99.5%, Extra Dry, stabilized, AcroSeal™
Cyclopentyl methyl ether, 99.5%, Extra Dry, stabilized, AcroSeal™
Cyclopentyl methyl ether, 99.5%, Extra Dry, stabilized, AcroSeal™
Cyclopentyl methyl ether, 99.5%, Extra Dry, stabilized, AcroSeal™
Cyclopentyl methyl ether, 99.5%, Extra Dry, stabilized, AcroSeal™
Acros Organics

Cyclopentyl methyl ether, 99.5%, Extra Dry, stabilized, AcroSeal™

CAS: 5614-37-9 | C6H12O | 100.16 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
1 L
Packaging:
AcroSeal™ Glass Bottle
Catalog number 397251000
also known as 39725
HSN 29092000
Price (INR)
6,237.00
Each
Add to cart
Quantity:
100 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (INR)
6,237.00
Each
Add to cart
Chemical Identifiers
CAS5614-37-9
IUPAC Namemethoxycyclopentane
Molecular FormulaC6H12O
InChI KeySKTCDJAMAYNROS-UHFFFAOYSA-N
SMILESCOC1CCCC1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Infrared spectrumConforms
Water=<0.005 %
Peroxide value=<50 ppm
GC>=99.4 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only