(R)-3-Amino-1,2-propanediol, 98%
(R)-3-Amino-1,2-propanediol, 98%
(R)-3-Amino-1,2-propanediol, 98%
Acros Organics

(R)-3-Amino-1,2-propanediol, 98%

CAS: 66211-46-9 | C3H9NO2 | 91.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 397880250
also known as 39788
HSN 29221990
Price (INR)
68,505.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
68,505.00
Each
Chemical Identifiers
CAS66211-46-9
IUPAC Name3-aminopropane-1,2-diol
Molecular FormulaC3H9NO2
InChI KeyKQIGMPWTAHJUMN-UHFFFAOYNA-N
SMILESNCC(O)CO
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point53°C to 57°C
Specific optical rotation+27° to +29° (20°C, 589 nm) (C=4, 5 N HCl)
Titration with HClO4>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only