Search
Search

| Catalog Number | Holds | For Use With (Equipment) |
|---|---|---|
| 398187 | Boxes (3 in.) | 13, 17, 20 cu. ft. Freezers |
| 114048 | General Freezer Items | 12.7, 20 cu. ft. Freezers |
| 189000 | General Freezer Items | 20 cu. ft. Freezers |
| 189001 | General Freezer Items | 12.7 cu. ft. Freezers |
| 398184 | Boxes (2 in.) | 3 cu. ft. Chest Freezers |
| 398185 | Boxes (3 in.) | 3 cu. ft. Freezers |
| 398188 | Plates | 12.7, 17, 20 cu. ft. Freezers |
Use Thermo Scientific™ Chest Freezer Racks with TDE Series Ultra-Low Temperature Chest Freezers as well as legacy Forma™, HERAfreeze™, Revco™ and TS-Series Ultra-Low Temperature Chest Freezers.