4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals
4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals
4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals
Acros Organics

4-Chloro-2-methoxybenzyl alcohol, 97%, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
1 g
Catalog number 398560050
also known as 39856
HSN 29072990
Price (INR)
15,742.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
15,742.00
Each
Chemical Identifiers
CAS90296-27-8
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to cream
Appearance (Form)Crystalline powder
Infrared spectrumConforms
GC>=96.0 %
Melting point47°C to 51°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only