L-allo-Isoleucine, 99%, 99% ee
L-allo-Isoleucine, 99%, 99% ee
L-allo-Isoleucine, 99%, 99% ee
Acros Organics

L-allo-Isoleucine, 99%, 99% ee

CAS: 1509-34-8 | C6H13NO2 | 131.18 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mg
1 g
Catalog number 399491000
also known as 39949
HSN 29224990
Price (INR)
21,872.00
Each
Quantity:
100 mg
Request bulk or custom format
Chemical Identifiers
CAS1509-34-8
IUPAC Name(2S,3R)-2-amino-3-methylpentanoic acid
Molecular FormulaC6H13NO2
InChI KeyAGPKZVBTJJNPAG-BJNJOQLTNA-N
SMILESCC[C@@H](C)[C@H](N)C(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
NMRconsistent with structure
Specific optical rotation+35° to +39° (20°C, 589 nm) (C=5, 1N HCl)
Enantiomeric excess>=98.5 % (HPLC)
Appearance (Color)White to off-white
Appearance (Form)Powder
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only