Mono-Ethyl malonate, 95%
Mono-Ethyl malonate, 95%
Mono-Ethyl malonate, 95%
Acros Organics

Mono-Ethyl malonate, 95%

CAS: 1071-46-1 | C5H8O4 | 132.11 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
5 g
Packaging:
Glass bottle
Catalog number 399900250
also known as 39990
HSN 29171990
Price (INR)
14,416.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
14,416.00
Each
Chemical Identifiers
CAS1071-46-1
IUPAC Name3-ethoxy-3-oxopropanoic acid
Molecular FormulaC5H8O4
InChI KeyHGINADPHJQTSKN-UHFFFAOYSA-N
SMILESCCOC(=O)CC(O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Liquid
Appearance (Color)Clear colorless to light yellow
Infrared spectrumConforms
Titration with NaOH>=94 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only