Printers for CryoMed™ and CryoPlus™ Freezers
Actual product may vary
Thermo Scientific™
Printers for CryoMed™ and CryoPlus™ Freezers
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with this optional printer.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeVoltage
4000566Thermal Printer PaperNot Applicable
4000565Thermal Data Printer120 V
4000665Thermal Data Printer220 V
Catalog number 4000566
Price (INR)
-
Type:
Thermal Printer Paper
Voltage:
Not Applicable
Enhance the functionality for Thermo Scientific™ CryoMed™ controlled-rate freezers with Thermo Scientific™ Thermal Printers for CryoMed and CryoPlus™ Freezers. These printers are available in 120V or 220V models, and include cable assembly and RS-232 connector kit.
  • Model 4000565 - 120V, 60Hz printer with cable assembly and RS-232 connector kit
  • Model 4000665 - 220V, 50Hz printer with cable assembly and RS-232 connector kit
  • Model 4000566 - Thermal printer paper (5 rolls/package)
  • Warranty varies by country, please contact us for details
Specifications
For Use With (Equipment)All CryoMed Controlled-Rate Freezers
TypeThermal Printer Paper
VoltageNot Applicable
Unit SizePack of 5