Product Image
Applied Biosystems™

Synthesis Column Filters

Have Questions?
Catalog NumberQuantity
400059100 each
Catalog number 400059
Price (INR)
0.01
Each
Quantity:
100 each
Price (INR)
0.01
Each
These filters are intended for use with empty synthesis columns to allow users to pack columns manually.

For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Oligo Synthesis
For Use With (Equipment)3900 DNA Synthesizer
Product TypeSynthesis Column Filter Set
Quantity100 each
Sample TypeDNA
Shipping ConditionRoom Temperature
Unit SizeEach
Contents & Storage
Store at Room Temperature. Includes 100 filters.