CryoMed™ Controlled-Rate Freezer IVF Accessories
CryoMed™ Controlled-Rate Freezer IVF Accessories
Thermo Scientific™
CryoMed™ Controlled-Rate Freezer IVF Accessories
Integrate Thermo Scientific™ CryoMed™ Controlled-Rate Freezer IVF Accessories with your current system for increased functionality.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
4000710Straw Holder
180255Sleeve Plug
4000711Vial Holder
4000714Cane Holder
Catalog number 4000710
Price (INR)
-
Request A Quote
Type:
Straw Holder
Integrate Thermo Scientific™ CryoMed™ Controlled-Rate Freezer IVF Accessories with your current system for increased functionality.
For use in CryoMed 7456, 7457, 7475, 7458, 7459, and 7476 freezers.
  • Warranty varies by country, please contact us for details
  • Specifications
    For Use With (Equipment)All CryoMed Controlled-Rate Freezers
    HoldsStraw
    TypeStraw Holder
    Unit SizeEach