CryoMed™ Controlled-Rate Freezer IVF Accessories
Thermo Scientific™
CryoMed™ Controlled-Rate Freezer IVF Accessories
Integrate Thermo Scientific™ CryoMed™ Controlled-Rate Freezer IVF Accessories with your current system for increased functionality.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberType
4000714Cane Holder
180255Sleeve Plug
4000710Straw Holder
4000711Vial Holder
Catalog number 4000714
Price (INR)
-
Type:
Cane Holder
Integrate Thermo Scientific™ CryoMed™ Controlled-Rate Freezer IVF Accessories with your current system for increased functionality.
For use in CryoMed 7456, 7457, 7475, 7458, 7459, and 7476 freezers.
  • Warranty varies by country, please contact us for details
  • Specifications
    Descriptionfor 1.2 and 2.0 vials
    For Use With (Equipment)All CryoMed Controlled-Rate Freezers
    HoldsCane
    TypeCane Holder
    Unit SizeEach