RapidHIT™ ID GlobalFiler™ Express Control (Ladder) 5-Cartridge Kit
Applied Biosystems™
RapidHIT™ ID GlobalFiler™ Express Control (Ladder) 5-Cartridge Kit
The Applied Biosystems RapidHIT ID GlobalFiler Express Control (Ladder) 5-Cartridge Kit is for use with the Applied Biosystems RapidHIT ID System.
Have Questions?
Catalog NumberQuantity
400096RH5 Cartridges
Catalog number 400096RH
Price (INR)
120,783.00
Each
Quantity:
5 Cartridges
Price (INR)
120,783.00
Each
The Applied Biosystems RapidHIT ID GlobalFiler Express Control (Ladder) 5-Cartridge Kit is for use with the Applied Biosystems RapidHIT ID System. This kit contains five ACE GlobalFiler Express Control (ladder) cartridges for use with ACE GlobalFiler Express Sample cartridges.
Required for use:
  • Applied Biosystems RapidHIT ID System (Cat. No. A41810)
  • Applied Biosystems RapidHIT ID Instrument Software v1.3.1 or higher
  • Applied Biosystems RapidHIT ID Primary Cartridge (Cat. No. A53084)
Specifications
For Use With (Application)Human Identification
For Use With (Equipment)RapidHIT™ ID System
Quantity5 Cartridges
FormatKit
Unit SizeEach