Product Image
Applied Biosystems™
CPG (Controlled Pore Glass) Synthesis Column, 500 Angstrom, 10.0 μmol, dG(iBu)
The 10.0 μmol dG(iBu) Controlled Pore Glass (CPG) columns are suitable for the synthesis of oligonucleotides <50 bases in length.
Have Questions?
Catalog NumberQuantity
4003921 Column
Catalog number 400392
Price (INR)
-
Quantity:
1 Column
The 10.0 μmol dG(iBu) Controlled Pore Glass (CPG) columns are suitable for the synthesis of oligonucleotides <50 bases in length. These columns deliver high-quality oligonucleotides.
  • Proprietary design and manufacturing methods ensure consistency and high quality
  • Each column is tested to verify accuracy and precision of fill, then pressure-tested to verify proper assembly and to guarantee each column is leak-free
  • The 500 angstrom pore size allows synthesis of oligonucleotides of up to 50 bases.
These columns are fully compatible with the Applied Biosystems 3400 instruments, ABI 391, ABI 392, ABI 394, and earlier instruments.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Column TypeOligonucleotide Synthesis
For Use With (Application)Oligo Synthesis
For Use With (Equipment)3900 DNA Synthesizer
High-throughput CompatibilityNot High-throughput Compatible (Manual)
MatrixControlled Pore Glass
Pore Size500 Å
Prep Scale10 μmol
Product TypeOligonucleotide Synthesis Columns
Quantity1 Column
Sample TypeOligos
Shipping ConditionRoom Temperature
TargetNucleic Acids
FormatColumn
Unit SizeEach
Contents & Storage
Columns may be stored in a dry environment at room temperature.