p-Anisidine hydrochloride, 99%
p-Anisidine hydrochloride, 99%
p-Anisidine hydrochloride, 99%
Acros Organics

p-Anisidine hydrochloride, 99%

CAS: 20265-97-8 | C7H9NO·HCl | 159.61 g/mol
Have Questions?
Catalog NumberQuantity
401300050
also known as 40130
5 g
Catalog number 401300050
also known as 40130
HSN 29222990
Price (INR)
4,401.00
Each
Quantity:
5 g
Request bulk or custom format
Price (INR)
4,401.00
Each
Chemical Identifiers
CAS20265-97-8
IUPAC Namehydrogen 4-methoxyaniline chloride
Molecular FormulaC7H10ClNO
InChI KeyVQYJLACQFYZHCO-UHFFFAOYSA-N
SMILES[H+].[Cl-].COC1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Off-white to pink to grey-brown
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Melting point214°C to 219°C
Titration with HClO4>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only