Iodine-Water-Pyridine-Tetrahydrofuran Oxidizer Solution (0.02 M)
This Iodine⁄Water⁄Pyridine⁄Tetrahydrofuran (oxidizer) solution comes in a convenient large volume for ABI 394 Nucleic Acid synthesizers, resulting in extended hands-freeRead more
Have Questions?
Change view
Catalog Number
Quantity
402191
2 L
401732
200 mL
401632
450 mL
4340867
4 x 180 mL
4340862
4 x 450 mL
4347380
4 x 2 L
6 Options
Catalog number 402191
Price (INR)
-
Quantity:
2 L
This Iodine⁄Water⁄Pyridine⁄Tetrahydrofuran (oxidizer) solution comes in a convenient large volume for ABI 394 Nucleic Acid synthesizers, resulting in extended hands-free DNA synthesis. With these larger bottles attached your synthesizer can produce up to 40 average primer-length oligonucleotides with no bottle changes, reducing attended operation time by 50%.
• Reagent concentrations are within our strict specifications, guaranteeing consistent quality. • Our performance specification of at least 98% stepwise coupling efficiency is assured, increasing oligonucleotide synthesis yields of full-length product. • Reagent kits are available for your convenience, and save you time and money.
Our high-performance reagents begin with raw materials that must conform to strict specifications. Once the reagents are manufactured and packaged, we perform extensive testing to assure quality. Reagent lifetime on the instrument is two weeks for tetrazole, and six weeks for all other reagents.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
Chemical Name or MaterialPyridine Tetrahydrofuran
For Use With (Application)Oligo Synthesis
For Use With (Equipment)3900 DNA Synthesizer
FormLiquid
Product TypeTetrahydrofuran Oxidizer Solution
Quantity2 L
Reagent TypeSolvent or General Reagent
Sample TypeDNA
Shipping ConditionRoom Temperature
FormatBottle
Unit SizeEach
Contents & Storage
Store in a cool, dry, and well-ventilated location, appropriate for flammable lliquids.