Bromophenol Blue, pure, 0.1% in water
Bromophenol Blue, pure, 0.1% in water
Bromophenol Blue, pure, 0.1% in water
Acros Organics

Bromophenol Blue, pure, 0.1% in water

CAS: 115-39-9 | C19H10Br4O5S | 669.96 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 mL
25 mL
Catalog number 403151000
HSN 28521000
Price (INR)
16,807.00
Each
Quantity:
100 mL
Request bulk or custom format
Price (INR)
16,807.00
Each
Chemical Identifiers
CAS115-39-9
IUPAC Name3,3-bis(3,5-dibromo-4-hydroxyphenyl)-3H-2,1λ⁶-benzoxathiole-1,1-dione
Molecular FormulaC19H10Br4O5S
InChI KeyUDSAIICHUKSCKT-UHFFFAOYSA-N
SMILESOC1=C(Br)C=C(C=C1Br)C1(OS(=O)(=O)C2=CC=CC=C12)C1=CC(Br)=C(O)C(Br)=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Concentration(by UV) About 0.1%
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Indicator
RUO – Research Use Only