2-Butoxyethyl acetate, 98%
2-Butoxyethyl acetate, 98%
2-Butoxyethyl acetate, 98%
Acros Organics

2-Butoxyethyl acetate, 98%

CAS: 112-07-2 | C8H16O3 | 160.21 g/mol
Have Questions?
Catalog NumberQuantity
403410100
also known as 40341
10 L
Catalog number 403410100
also known as 40341
HSN 29153990
Price (INR)
46,112.00
Each
Quantity:
10 L
Request bulk or custom format
Price (INR)
46,112.00
Each
Chemical Identifiers
CAS112-07-2
SpecificationsSpecification SheetSpecification Sheet
GC>=97.5 %
Appearance (Form)Clear liquid
Infrared spectrumConforms
Refractive index1.4120 to 1.4150 (20°C, 589 nm)
Color scale=<15 APHA
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only