Deoxyribonucleic acid sodium salt, pure, M.W.  50,000 to 100,000 Dalton
Deoxyribonucleic acid sodium salt, pure, M.W.  50,000 to 100,000 Dalton
Deoxyribonucleic acid sodium salt, pure, M.W.  50,000 to 100,000 Dalton
Acros Organics

Deoxyribonucleic acid sodium salt, pure, M.W. 50,000 to 100,000 Dalton

CAS: 68938-01-2 | C17H15BrFNO3 | 50000-100000 g/mol
Have Questions?
Catalog NumberQuantityPackaging
406251000
also known as 40625
100 gGlass bottle
Catalog number 406251000
also known as 40625
HSN 29349900
Price (INR)
37,898.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
37,898.00
Each
Chemical Identifiers
CAS68938-01-2
IUPAC NameDeoxyribonucleic acid sodium salt
SMILES*
Molecular Weight (g/mol)50000-100000
SynonymDNA sodium salt
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Almost white to light yellow to tan
Appearance (Form)Powder
Phosphorus (P)=<9 %
Infrared spectrumConforms
Loss on drying=<18 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only