trans-1,4-Dibromo-2-butene, 97%
trans-1,4-Dibromo-2-butene, 97%
trans-1,4-Dibromo-2-butene, 97%
Acros Organics

trans-1,4-Dibromo-2-butene, 97%

CAS: 821-06-7 | C4H6Br2 | 213.9 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 406460250
also known as 40646
HSN 29033919
Price (INR)
9,114.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
9,114.00
Each
Chemical Identifiers
CAS821-06-7
IUPAC Name(2E)-1,4-dibromobut-2-ene
Molecular FormulaC4H6Br2
InChI KeyRMXLHIUHKIVPAB-OWOJBTEDSA-N
SMILESBrCC=CCBr
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to beige to brown
Appearance (Form)Crystalline powder or crystals
GC>=96.0 %
Infrared spectrumConforms
Melting point48°C to 53°C
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only