Didodecyldimethylammonium bromide, 99%
Didodecyldimethylammonium bromide, 99%
Didodecyldimethylammonium bromide, 99%
Acros Organics

Didodecyldimethylammonium bromide, 99%

CAS: 3282-73-3 | C26H56BrN | 462.64 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
25 g
Packaging:
Plastic bottle
Catalog number 407121000
also known as 40712
HSN 29215990
Price (INR)
53,640.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
53,640.00
Each
Chemical Identifiers
CAS3282-73-3
IUPAC Namedidodecyldimethylazanium bromide
Molecular FormulaC26H56BrN
InChI KeyXRWMGCFJVKDVMD-UHFFFAOYSA-M
SMILES[Br-].CCCCCCCCCCCC[N+](C)(C)CCCCCCCCCCCC
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Melting point158°C to 163°C
Titration Argentometric>=98.5 %
Appearance (Color)White to off-white
Appearance (Form)Powder or crystals
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Phase transfer catalyst
RUO – Research Use Only