Product Image
Gibco™

Basal Medium Eagle (BME)

Basal Medium Eagle (BME) is a widely used synthetic basal medium for supporting the growth of many different mammalian cells.Read more
Have Questions?
Change viewbuttonViewtableView
Catalog NumberQuantity
41010026
also known as 41010-026
500 mL
41010109
also known as 41010-109
10 x 500 mL
Catalog number 41010026
also known as 41010-026
Price (INR)
-
Quantity:
500 mL
Customize this product

Basal Medium Eagle (BME) is a widely used synthetic basal medium for supporting the growth of many different mammalian cells. BME was originally developed by Harry Eagle for HeLa cells and mouse fibroblasts, when he discovered the minimum requirements for cell growth in vitro. Gibco™ BME has since been used for other human lines, including WI-38 and MRC-5.

This BME is modified as follows:
WithWithout
• Phenol Red• L-glutamine
 • HEPES


The complete formulation is available.

Gibco™ BME contains eight B vitamins, the ten essential amino acids, plus cystine, tyrosine, and glutamine. There are a number of modifications of BME: Minimal Essential Medium, Glasgow's Medium and Dulbecco's Modified Eagle's Medium. This BME formulation contains Earle's salts for use in a CO2 incubator.

Fetal Bovine Serum (FBS). BME uses a sodium bicarbonate buffer system (2.2 g/L) and therefore requires a 5-10% CO2 environment to maintain physiological pH.

For Research Use or Further Manufacturing. Not for diagnostic use or direct administration into humans or animals.
Specifications
Concentration1 X
Manufacturing QualitycGMP-compliant under the ISO 13485 standard
Product LineGibco
Product TypeBasal Medium Eagle (BME)
Quantity500 mL
Shelf Life12 Months
Shipping ConditionRoom Temperature
Culture TypeMammalian Cell Culture
FormLiquid
SterilitySterile-filtered
With AdditivesLow Glucose, Phenol Red
Without AdditivesNo Glutamine, No HEPES, No Sodium Pyruvate
Unit SizeEach
Contents & Storage
Storage conditions: 2-8°C. Protect from light
Shipping conditions: Ambient
Shelf life: 12 months from date of manufacture