Barnstead™ MegaPure™ Bottles
Actual product may vary
Barnstead™ MegaPure™ Bottles
Barnstead™ MegaPure™ Bottles
Barnstead™ MegaPure™ Bottles
Barnstead™ MegaPure™ Bottles
Thermo Scientific™
Barnstead™ MegaPure™ Bottles
Safely store distilled water using plastic or glass Thermo Scientific™ Barnstead™ MegaPure™ Bottles compatible with Barnstead Mega-Pure Glass Stills.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Material
410164TS
also known as 410164
45 LGlass
41393413 LGlass
4139646 LPolyethylene
Catalog number 410164TS
also known as 410164
Price (INR)
-
Capacity (Metric):
45 L
Material:
Glass
Safely store distilled water using plastic or glass Thermo Scientific™ Barnstead™ MegaPure™ Bottles compatible with Barnstead Mega-Pure Glass Stills. Water bottles for this distillation system are available in varying capacities.
Specifications
Capacity (English)11.2 gal.
Capacity (Metric)45 L
MaterialGlass
For Use With (Equipment)MegaPure Glass Still
TypeGlass
Unit SizeEach