Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks
Thermo Scientific™

Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks

Combine these flasks with most lab reagents and solutions due to their excellent chemical compatibility and shatter-proof construction.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberCapacity (Metric)Closure Size
4102-0250PK
also known as 4102-0250
250 mLStopper No. 6
4102-0125PK
also known as 4102-0125
125 mLStopper No. 3
Catalog number 4102-0250PK
also known as 4102-0250
Price (INR)
9,314.00
Pack of 6
Capacity (Metric):
250 mL
Closure Size:
Stopper No. 6
Price (INR)
9,314.00
Pack of 6

Choose Thermo Scientific™ Nalgene™ Polypropylene Copolymer Erlenmeyer Flasks for use with a wide range of lab reagents and solutions due to their excellent chemical compatibility. Rugged, shatterproof construction and light weight compared with glass make these flasks a valuble element of lab safety programs.

  • Translucent wall makes liquid level easily visible
  • 2000mL size has molded-in graduations
  • Mouth sized to accept standard sized stoppers (stopper sold separately)
  • Repeatedly autoclavable for long life and rigorous use

Recycling Information:
Nalgene reusable labware products are designed to last many years under typical laboratory conditions. Yet, as with all durable products, they need to be retired and replaced eventually. Thermo Fisher Scientific encourages recycling Nalgene polypropylene (PP) and polypropylene copolymer (PPCO) labware at end of product life in communities where polypropylene can be recycled. Please also recycle the corrugate cartons and any plastic bags (LDPE) included in your product packaging.

Specifications
Capacity (English)8 oz.
Closure SizeStopper No. 6
DescriptionFlask Erlenmeyer PP 250mL
No. per Case12
No. per Pack6
AutoclavableAutoclavable
Bottom ShapePlain
Capacity (Metric)250 mL
DisposableReusable
GraduatedNo
MaterialPPCO
Mouth StyleNarrow
SterilityNon-sterile
Unit SizePack of 6