Product Image
Applied Biosystems™
Fmoc-L-Arginine (Mtr), 10/Pk.
These are Amino Acid Cartridges for the ABI™ 431-433 Peptide Synthesizer.For Research Use Only. Not for use in diagnostics procedures.Read more
Have Questions?
Catalog NumberQuantity
41063310 Ea.
Catalog number 410633
Price (INR)
-
Quantity:
10 Ea.
These are Amino Acid Cartridges for the ABI™ 431-433 Peptide Synthesizer.
For Research Use Only. Not for use in diagnostics procedures.
For Research Use Only. Not for use in diagnostic procedures.
Specifications
For Use With (Application)Protein Sequencing & Peptide Synthesis
For Use With (Equipment)433 A Peptide Synthesizer
Product TypePeptide Synthesis Reagents
Quantity10 Ea.
FormatCartridge
Unit SizeEach
Contents & Storage
Each package contains 10 cartridges.