Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks
Actual product may vary
Thermo Scientific™

Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks

Ensure a sterile gas exchange for shaker and suspension cell culture by using Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberFor Use With (Equipment)
4114-0045500, 1000, 2000mL flasks
4114-0038125, 250mL flasks
Catalog number 4114-0045
Price (INR)
5,961.00
Case of 12
For Use With (Equipment):
500, 1000, 2000mL flasks

Ensure a sterile gas exchange for shaker and suspension cell culture by using Thermo Scientific™ Nalgene™ Vented HDPE Closures for Sterile Single Use Erlenmeyer Flasks.

  • Blue high-density polyethylene with hydrophobic 0.2μm PTFE membrane
  • Air exchange without unscrewing closure
  • Membrane heat welded into place for effective sealing
Warranty: 90 days
Disclaimer to internal research use restriction: Any restrictions on the purchaser to utilize the product(s) for internal research purposes only does not apply to the product(s) on the web pages associated with this statement.
Specifications
Closure TypeVented
ColorBlue
For Use With (Equipment)500, 1000, 2000mL flasks
No. per Case12
No. per Pack1
MaterialHDPE
SterilitySterile
Unit SizeCase of 12