Safely store distilled water using plastic or glass Thermo Scientific™ Barnstead™ MegaPure™ Bottles compatible with Barnstead Mega-Pure Glass Stills.
Have Questions?
Change view
Catalog Number
Capacity (Metric)
Material
413934
13 L
Glass
410164TS
also known as 410164
45 L
Glass
413964
6 L
Polyethylene
3 Options
Catalog number 413934
Price (INR)
-
Capacity (Metric):
13 L
Material:
Glass
Safely store distilled water using plastic or glass Thermo Scientific™ Barnstead™ MegaPure™ Bottles compatible with Barnstead Mega-Pure Glass Stills. Water bottles for this distillation system are available in varying capacities.