Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals
Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals
Acros Organics

Methyl sulfoxide, 99.9+%, ACS reagent, Thermo Scientific Chemicals

Have Questions?
Change viewbuttonViewtableView
Quantity:
1 L
500 mL
2.5 L
Packaging:
View all
Plastic Bottle
Catalog number 414880010
also known as 41488
HSN 29041090
Price (INR)
16,120.00
Each
Quantity:
1 L
Packaging:
Plastic Bottle
Request bulk or custom format
Price (INR)
16,120.00
Each
Chemical Identifiers
CAS67-68-5
IUPAC Namemethanesulfinylmethane
Molecular FormulaC2H6OS
InChI KeyIAZDPXIOMUYVGZ-UHFFFAOYSA-N
SMILESCS(C)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless
GC>=99.9 %
Water=<0.1 % (Coulometric)
Residue after evaporation=<0.01 %
Titrable acid=<0.001 meq/g
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only