1,2,2,6,6-Pentamethylpiperidine, 95%
1,2,2,6,6-Pentamethylpiperidine, 95%
1,2,2,6,6-Pentamethylpiperidine, 95%
Acros Organics

1,2,2,6,6-Pentamethylpiperidine, 95%

CAS: 79-55-0 | C10H21N | 155.28 g/mol
Have Questions?
Catalog NumberQuantityPackaging
416960050
also known as 41696
5 gGlass bottle
Catalog number 416960050
also known as 41696
HSN 29333919
Price (INR)
12,783.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
12,783.00
Each
Chemical Identifiers
CAS79-55-0
IUPAC Name1,2,2,6,6-pentamethylpiperidin-1-ium
Molecular FormulaC10H22N
InChI KeyXULIXFLCVXWHRF-UHFFFAOYSA-O
SMILESC[NH+]1C(C)(C)CCCC1(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Clear colorless to light yellow
Appearance (Form)Liquid
Infrared spectrumConforms
GC>=94 %
Refractive index1.4570 to 1.4600 (20°C, 589 nm)
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only