Pepsin, for biochemistry, powder
Acros Organics

Pepsin, for biochemistry, powder

Pepsin from porcine gastric mucosa, CAS # 9001-75-6, is a peptidase that is commonly used in enzymatic digestion of proteins. | CAS: 9001-75-6
Have Questions?
Catalog NumberQuantity
41707020020 g
Catalog number 417070200
HSN 98020000
Price (INR)
6,297.00
Each
Quantity:
20 g
Request bulk or custom format
Price (INR)
6,297.00
Each
Chemical Identifiers
CAS9001-75-6
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White to light yellow
Appearance (Form)Powder
Infrared spectrumConforms
Assay>=0.5 Ph. Eur. U/mg (Activity)
AssayHemoglobin, pH 1.6, 25°C
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

General Description

  • Pepsin is an endopeptidase that digests proteins by hydrolyzing peptide bonds
  • It is the active form of pepsinogen, which is produced in the chief cells of the gastric mucosa

Applications

  • Pepsin is commonly used to produce F(ab’)2 fragments from antibodies
  • It has been used along with other enzymes to demonstrate the effects of fixation and enzymatic digestion in immunohistochemical assays, using paraffin-embedded tissue
  • It can be used in hydrolyzing dry cervical samples in mice
  • It has been used in the digestion of crude wheat gliadin
  • It has been used to stimulate in vitro gastric digestion of cooked cod, cocoa mass, and dietary fiber
RUO – Research Use Only