Poly(ethylene glycol), average M.W. 1450
Poly(ethylene glycol), average M.W. 1450
Poly(ethylene glycol), average M.W. 1450
Acros Organics

Poly(ethylene glycol), average M.W. 1450

CAS: 25322-68-3 | (C2H4O)n | 62.07 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
100 g
1 kg
3 kg
Packaging:
View all
Plastic bottle
Catalog number 418045000
also known as 41804
HSN 98020000
Price (INR)
3,857.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
3,857.00
Each
Chemical Identifiers
CAS25322-68-3
Molecular Formula(C2H4O)n
InChI KeyLYCAIKOWRPUZTN-UHFFFAOYSA-N
SMILES[H]OCCO
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Waxy flakes
Infrared spectrumConforms
Water=<0.5 % (K.F.)
Hydroxyl value70 to 80 mg KOH/g
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only