Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™
Acros Organics

Potassium bis(trimethylsilyl)amide, 0.7M (15 wt.%) solution in toluene, AcroSeal™

CAS: 40949-94-8 | C6H18KNSi2 | 199.485 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
800 mL
100 mL
Packaging:
View all
AcroSeal™ Glass Bottle
Catalog number 418238000
HSN 29319090
Price (INR)
69,855.00
Each
Quantity:
800 mL
Packaging:
AcroSeal™ Glass Bottle
Request bulk or custom format
Price (INR)
69,855.00
Each
Chemical Identifiers
CAS40949-94-8
IUPAC Namepotassium bis(trimethylsilyl)azanide
Molecular FormulaC6H18KNSi2
InChI KeyIUBQJLUDMLPAGT-UHFFFAOYSA-N
SMILES[K+].C[Si](C)(C)[N-][Si](C)(C)C
View more
SpecificationsSpecification SheetSpecification Sheet
Titration with HClO40.6 to 0.9 M
Appearance (Form)Solution
Appearance (Color)Clear to slightly hazy colorless to amber
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only