Quinhydrone, 98%
Quinhydrone, 98%
Quinhydrone, 98%
Acros Organics

Quinhydrone, 98%

Quinhydrone, 98%, Quantity: 100g, Packaging: Glass bottle, Green, Melting Point: 168 deg.C to 172 deg.C, Molecular Weight: 218.21, Percent Purity: 98%, Assay Percent Range: 48.0 to 52% Hydroquinone (HPLC), CAS: 106-34-3, ChEBI: CHEBI:26491, Density: 1.4000g/mL | CAS: 106-34-3 | C12H10O4 | 218.21 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
Packaging:
Glass bottle
Catalog number 418770250
also known as 41877
HSN 29420011
Price (INR)
4,640.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,640.00
Each
Chemical Identifiers
CAS106-34-3
IUPAC Namebenzene-1,4-diol; cyclohexa-2,5-diene-1,4-dione
Molecular FormulaC12H10O4
InChI KeyBDJXVNRFAQSMAA-UHFFFAOYSA-N
SMILESOC1=CC=C(O)C=C1.O=C1C=CC(=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Infrared spectrumConforms
Iron (Fe)=<10 ppm
Sulfated ash=<0.2 %
HPLC48.0 to 52.0 % p-Benzoquinone
Titration Iodometric>=97.5 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only