Basic Fuchsin, 88%, pure
Basic Fuchsin, 88%, pure
Basic Fuchsin, 88%, pure
Acros Organics

Basic Fuchsin, 88%, pure

CAS: 632-99-5 | C20H20ClN3 | 337.85 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 419141000
HSN 32041329
Price (INR)
32,501.00
Each
Quantity:
100 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
32,501.00
Each
Chemical Identifiers
CAS632-99-5
IUPAC Namehydrogen 4-[(4-aminophenyl)(4-imino-3-methylcyclohexa-2,5-dien-1-ylidene)methyl]aniline chloride
Molecular FormulaC20H20ClN3
InChI KeyAXDJCCTWPBKUKL-UHFFFAOYSA-N
SMILES[H+].[Cl-].CC1=CC(C=CC1=N)=C(C1=CC=C(N)C=C1)C1=CC=C(N)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Green
Appearance (Form)Powder
Loss on drying=<10 % (105°C)
Infrared spectrumConforms
Dye content>=83 % (on dry substance)
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only