1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline), 99%
1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline), 99%
1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline), 99%
Acros Organics

1',3',3'-Trimethyl-6-hydroxyspiro(2H-1-benzopyran-2,2'-indoline), 99%

CAS: 23001-29-8 | C19H19NO2 | 293.37 g/mol
Have Questions?
Catalog NumberQuantityPackaging
421920050
also known as 42192
5 gGlass bottle
Catalog number 421920050
also known as 42192
HSN 32049000
Price (INR)
26,039.00
Each
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
26,039.00
Each
Chemical Identifiers
CAS23001-29-8
IUPAC Name1',3',3'-trimethyl-1',3'-dihydrospiro[chromene-2,2'-indol]-6-ol
Molecular FormulaC19H19NO2
InChI KeyUHRBDHPBCHWWAG-UHFFFAOYNA-N
SMILESCN1C2=CC=CC=C2C(C)(C)C11OC2=CC=C(O)C=C2C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light brown to purple
Infrared spectrumConforms
Appearance (Form)Powder
HPLC>=98.5 %
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only