Temperature Control Connectivity Accessories
Thermo Scientific™
Temperature Control Connectivity Accessories
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
Have Questions?
Change viewbuttonViewtableView
Catalog NumberTypeFor Use With (Equipment)
422000000004SoftwareRS-232 Communications Interface
16000331.8 m Interface CableCold/Heated Bath Circulators
16000345 ft. Interface CableCold/Heated Bath Circulators
16000355 m Interface CableCold/Heated Bath Circulators
42600000004425 ft. Communication CableChillers with RS232/RS485
160-0076Communication Extension BoardRecirculating Chiller or Bath
413000000005RTD Probe with 12 ft. cableThermoFlex Chillers
126000000024RTD Probe with 30 ft. cableThermoFlex Chillers
126000000052RTD Probe with 50 ft. cableThermoFlex Chillers
Catalog number 422000000004
Price (INR)
-
Type:
Software
For Use With (Equipment):
RS-232 Communications Interface
Maximize the efficiency of your temperature control device with Thermo Scientific™ Temperature Control Connectivity and monitoring accessories.
Connect your Thermo Scientific temperature control solution with your application.
  • Available for temperature control devices equipped with communications
  • Designed to work with your Thermo Scientific recirculating chiller or bath equipped with communications
  • Available in popular configurations that match your device’s connections and interface with your process
  • Take the guess work out of finding parts to match your recirculating chiller or bath
Thermo Scientific™ NEScom™ Windows-based software
  • Provides remote control of Thermo Scientific units equipped with an RS-232 communications interface
  • Choose upper and lower temperature alarm limits and monitor system status with an alarm
  • Record your results on a user selectable graph
  • Select from easy–to–use product icons
  • Create graphs and charts
  • Easily configure ramping functions over time
  • View pop-up alarm windows which display the error condition
  • View a virtual controller screen
  • PC compatible only
Specifications
CommunicationsRS232
For Use With (Equipment)RS-232 Communications Interface
TypeSoftware
Unit SizeEach