Copper(II) sulfate, 98%, pure, anhydrous
Copper(II) sulfate, 98%, pure, anhydrous
Copper(II) sulfate, 98%, pure, anhydrous
Acros Organics

Copper(II) sulfate, 98%, pure, anhydrous

CAS: 7758-98-7 | CuO4S | 159.6 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
5 g
100 g
500 g
2.5 kg
Packaging:
Glass bottle
Catalog number 422870050
HSN 28332500
Quantity:
5 g
Packaging:
Glass bottle
Request bulk or custom format
Chemical Identifiers
CAS7758-98-7
IUPAC Namecopper(2+) sulfate
Molecular FormulaCuO4S
InChI KeyARUVKPQLZAKDPS-UHFFFAOYSA-L
SMILES[Cu++].[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Powder
Titration Iodometric>=97.5 %
Chloride (Cl)=<50 ppm
Iron (Fe)=<100 ppm
Appearance (Color)Light grey
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only