Barium acetate, 99+%, ACS reagent
Barium acetate, 99+%, ACS reagent
Barium acetate, 99+%, ACS reagent
Acros Organics

Barium acetate, 99+%, ACS reagent

CAS: 543-80-6 | C4H6BaO4 | 255.42 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
100 g
500 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 423431000
HSN 29152990
Price (INR)
4,166.00
Each
Quantity:
100 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
4,166.00
Each
Chemical Identifiers
CAS543-80-6
IUPAC Namebarium(2+) diacetate
Molecular FormulaC4H6BaO4
InChI KeyITHZDDVSAWDQPZ-UHFFFAOYSA-L
SMILES[Ba++].CC([O-])=O.CC([O-])=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)White
Appearance (Form)Crystalline powder
Titration Complexometric99.0 to 102.0 %
Heavy metals (ICP-OES)=<5 ppm
Insoluble matter=<0.01 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only