4-Dimethylaminobenzaldehyde, ACS reagent
4-Dimethylaminobenzaldehyde, ACS reagent
4-Dimethylaminobenzaldehyde, ACS reagent
Acros Organics

4-Dimethylaminobenzaldehyde, ACS reagent

CAS: 100-10-7 | C9H11NO | 149.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
25 g
100 g
500 g
2.5 kg
Packaging:
View all
Glass bottle
Catalog number 423630250
also known as 42363
HSN 29223900
Price (INR)
4,261.00
Each
Quantity:
25 g
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
4,261.00
Each
Chemical Identifiers
CAS100-10-7
IUPAC Name4-(dimethylamino)benzaldehyde
Molecular FormulaC9H11NO
InChI KeyBGNGWHSBYQYVRX-UHFFFAOYSA-N
SMILESCN(C)C1=CC=C(C=O)C=C1
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Color)Light yellow to light green
Appearance (Form)Crystalline powder
Infrared spectrumConforms
Residue after ignition=<0.1 %
Solubility(in alcohol) Passes test
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only