Ammonium iron(III) sulfate dodecahydrate, 99%, ACS reagent
Ammonium iron(III) sulfate dodecahydrate, 99%, ACS reagent
Ammonium iron(III) sulfate dodecahydrate, 99%, ACS reagent
Acros Organics

Ammonium iron(III) sulfate dodecahydrate, 99%, ACS reagent

CAS: 7783-83-7 | H4FeNO8S2·12H2O | 482.19 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 g
25 g
2.5 kg
Packaging:
Plastic bottle
Catalog number 423695000
HSN 28332990
Price (INR)
15,073.00
Each
Quantity:
500 g
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
15,073.00
Each
Chemical Identifiers
CAS7783-83-7
IUPAC Nameiron(3+) ammonium dodecahydrate disulfate
Molecular FormulaFeH28NO20S2
InChI KeyLCPUDZUWZDSKMX-UHFFFAOYSA-K
SMILES[NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[Fe+3].[O-]S([O-])(=O)=O.[O-]S([O-])(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Adhering powder or crystals
Appearance (Color)Violet
Titration Iodometric98.5 to 102.0 %
Ferrous iron (Fe 2+)(Limit about 0.001%) Passes test
Magnesium (Mg)=<0.005 %
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only