Hydrochloric acid, ACS reagent, ca. 37% solution in water
Hydrochloric acid, ACS reagent, ca. 37% solution in water
Hydrochloric acid, ACS reagent, ca. 37% solution in water
Acros Organics

Hydrochloric acid, ACS reagent, ca. 37% solution in water

CAS: 7647-01-0 | ClH | 36.46 g/mol
Have Questions?
Change viewbuttonViewtableView
Quantity:
500 mL
2.5 L
25 L
Packaging:
View all
Plastic bottle
Glass Bottle
Catalog number 423795001
HSN 28061000
Price (INR)
4,607.00
Each
Quantity:
500 mL
Packaging:
Plastic bottle
Request bulk or custom format
Price (INR)
4,607.00
Each
Chemical Identifiers
CAS7647-01-0
IUPAC Namehydrogen chloride
Molecular FormulaClH
InChI KeyVEXZGXHMUGYJMC-UHFFFAOYSA-N
SMILESCl
View more
SpecificationsSpecification SheetSpecification Sheet
Residue after ignition=<0.0005 %
Sulfite (SO3)=<1 ppm
Iron (Fe)=<0.2 ppm
Ammonium (NH4)=<3 ppm
Arsenic (As)=<0.01 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
RUO – Research Use Only