Perchloric acid, ACS reagent, ca. 70% solution in water
Perchloric acid, ACS reagent, ca. 70% solution in water
Perchloric acid, ACS reagent, ca. 70% solution in water
Perchloric acid, ACS reagent, ca. 70% solution in water
Acros Organics

Perchloric acid, ACS reagent, ca. 70% solution in water

CAS: 7601-90-3 | ClHO4 | 100.45 g/mol
Have Questions?
Catalog NumberQuantityPackaging
424030025
also known as 42403
2.5 kgGlass bottle
Catalog number 424030025
also known as 42403
HSN 28439019
Price (INR)
35,933.00
Each
Add to cart
Quantity:
2.5 kg
Packaging:
Glass bottle
Request bulk or custom format
Price (INR)
35,933.00
Each
Add to cart
Chemical Identifiers
CAS7601-90-3
IUPAC Nameperchloric acid
Molecular FormulaClHO4
InChI KeyVLTRZXGMWDSKGL-UHFFFAOYSA-N
SMILESO[Cl](=O)(=O)=O
View more
SpecificationsSpecification SheetSpecification Sheet
Appearance (Form)Clear liquid
Sulfate (SO4)=<10 ppm
Color scale=<10 APHA
Titration with NaOH69.0 to 72.0 %
Heavy metals (as Pb)=<1 ppm
View more
This Thermo Scientific Chemicals brand product was originally part of the Acros Organics product portfolio. Some documentation and label information may refer to the legacy brand. The original Acros Organics product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Oxidation reagent
RUO – Research Use Only